This product is now discontinued, View Related Products
Memmert General-Purpose Mechanical Convection Ovens
Precise drying and heating in research, science, industry and quality assurance.
  • Quick and intuitive operation through the unique ControlCOCKPIT
  • AtmoCONTROL software provides simple drag-and-drop programming and documentation
  • Scratch-resistant structured stainless steel exterior
  • Maintain records with 10-year internal data logger and Ethernet interface
These mechanical convection ovens are perfect for heating and drying applications in research or industrial environments. Featuring the Memmert ControlCOCKPIT for quick and intuitive control with simple touch, turn and go operation. Durable construction with a scratch-resistant structured stainless steel exterior and easy-to-clean corrosion-resistant stainless steel interior. Recessed grips make oven easy to carry and handle and stainless steel door can be changed from left to right side opening. Other features include an internal data logger with a storage capacity of 10 years and an Ethernet interface for uploading and implementing programs and for online logging.
635,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Memmert UF 30 230 VOLT Universal Mechanical Oven, 1.1 Cuft, Single Display, 230V WW-52200-19 Mfr # UF 30 230 VOLT
COMPARE
1.1 230
₹635,000.00INR / Each
Memmert UF 75 PLUS 115 VOLT Universal Mechanical Oven, Twin Display, 2.7 Cuft, 115V WW-52200-32 Mfr # UF 75 PLUS 115 VOLT
COMPARE
2.6 115