Meiji Professional Compound Microscopes
Semi-plan objectives give high-image quality, contrast, and flatness of field
  • Koehler illumination provides an evenly illuminated image without reflection or glare
  • Smooth-operating, ball-bearing quintuple nosepiece provides effortless objective changes
These professional microscopes feature a fully rotatable, 30°> inclined binocular head, synchronized interpupillary adjustment from 53 mm to 72 mm, and diopter adjustment for each ocular tube.
Feature semi-plan 4x, 10x, 40x, and 100x objectives that flatten out your field of view and give you sharper images. Quintuple reverse-angle nosepiece gives you easy access to the specimen and stage. The smooth operating nosepiece has an extra hole to accommodate a supplementary objective.
All models also feature paired, DIN wide-field 10x eyepieces. Microscopes have coaxial coarse and fine focus controls; coarse focus has tension adjustment to accommodate for each individual user.
526,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Maximum Magnification Power (VAC)
Availability Pricing
Meiji Techno ML5000 Compound Binocular Microscope; 40/100/400/1000x, 30W, Semi-Plan, 220V WW-48401-25 Mfr # ML5000/220VAC
COMPARE
Binocular 1000x 220
₹526,000.00INR / Each