Meiji Techno Microscope Eyepieces
Interchangeable eyepieces enhance flexibility of microscopes
  • Compatible with Meiji Techno EMZ, EMT, and EMF model microscopes
These paired interchangeable widefield eyepieces provide added customization to the Meiji Techno EMZ, EMT, and EMF model microscopes. Easily change eyepiece magnification for a wide range of applications. Microscopes sold separately.
41,400.00 - ₹53,800.00INR / Pair of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Eyepiece Magnification Field of View Description
Availability Pricing
Meiji Techno Microscope Eyepieces, Super Widefield, 10X, 23 mm Field of View; Pair WW-48402-58 Mfr # MA502
COMPARE
10x 23 mm Microscope Eyepieces, Super Widefield, 10X, 23 mm Field of View; Pair
₹41,400.00INR / Pair of 1
Meiji Techno Microscope Eyepieces, Super Widefield, 15X, 15.4 mm Field of View; Pair WW-48402-60 Mfr # MA503
COMPARE
15x 15.4 mm Microscope Eyepieces, Super Widefield, 15X, 15.4 mm Field of View; Pair
₹41,400.00INR / Pair of 1
Meiji Techno Microscope Eyepieces, Super Widefield, 20X, 11.4 mm Field of View; Pair WW-48402-62 Mfr # MA504
COMPARE
20x 11.5 mm Microscope Eyepieces, Super Widefield, 20X, 11.4 mm Field of View; Pair
₹53,800.00INR / Pair of 1