This product is now discontinued, View Related Products
Meiji Metallurgical Microscopes
View metals, plastics, ceramics, paint, glass, crystals, and semiconductor components
  • Plan Epi optics provide clear images, excellent color rendering, and superior flatness of field
Metallurgical microscopes illuminate the specimen through the objective lens — ideal for specimens that don't transmit light, such as metals. Comfortable Siedentopf viewing heads have 30'' inclined eyetubes with one adjustable diopter and one fixed diopter. The interpupillary distance adjustable from 53 to 75 mm. Ergonomic coaxial focus controls have tension adjustment and a safety autostop. Koehler illumination allows for effective contrast control. The illuminator features field iris, aperature iris, and filter slots. Filter sets are included.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Head Configuration Illuminator(s)
Availability Pricing
Meiji Techno MX7000 Metallurgical Binocular Microscope; Brightfield/Incident WW-48405-21 Mfr # MT7000
COMPARE
Binocular Brightfield/Incident
Call For Price
Meiji Techno MX7100 Metallurgical Trinocular Microscope; Brightfield/Incident WW-48405-27 Mfr # MT7100
COMPARE
Trinocular Brightfield/Incident
Call For Price
Meiji Techno MX7520 Metallurgical Binocular Microscope; Brightfield/Darkfield/Incident WW-48405-31 Mfr # MT7520
COMPARE
Binocular Brightfield/Darkfield/Incident
Call For Price
Meiji Techno MX7530 Metallurgical Trinocular Microscope; Brightfield/Darkfield/Incident WW-48405-37 Mfr # MT7530
COMPARE
Trinocular Brightfield/Darkfield/Incident
Call For Price
Meiji Techno MX8000 Metallurgical Binocular Microscope; Brightfield/Incident/Transmitted WW-48405-41 Mfr # MT8000
COMPARE
Binocular Brightfield / Incident / Transmitted
Call For Price
Meiji Techno MX8100 Metallurgical Trinocular Microscope; Brightfield/Incident/Transmitted WW-48405-47 Mfr # MT8100
COMPARE
Trinocular Brightfield / Incident / Transmitted