This product is now discontinued, View Related Products
Meiji Epi-Fluorescent Microscopes
Semi Apo Plan objectives provide bright, high-contrast images, with UV transmission and color correction
Epi-fluorescent microscope is designed for use with specimens that self fluoresce, or that use a fluorescent dye to bring out particular properties of the specimen. Ideal for use in cell biology and laboratory research. Modular six-position epi-fluorescent illuminator features six blank positions for filter cubes; basic blue, green, and UV-excitation filter sets mounted in filter cubes are included. Lamp house uses a 100 W mercury arc lamp.
Microscope features include siedentopf head for adjustability, diopter adjustment on one eyepiece, ceramic-coated stage, Abbe condenser with adjustable field iris, bottom halogen illumination for transmitted light use, and ergonomically placed focus controls.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Head Configuration
Availability Pricing
Meiji Techno Epi-Fluorescent Microscope, Binocular WW-48924-60 Mfr # MT6200H
COMPARE
Binocular
Call For Price
Meiji Techno Epi-Fluorescent Microscope, Trinocular WW-48924-65 Mfr # MT6300H
COMPARE
Trinocular