Meiji Techno Microscope C-Mount Adapters
Easily connect your Meiji Techno microscope to a camera
  • Compatible with Meiji Techno microscopes
Use C-mount adapters to attach a digital camera or video camera to the photo tube of your Meiji Techno microscope. The C-mount adapter that is needed will depend on your microscope, as well as your camera. Camera and microscope sold separately.
35,200.00 - ₹73,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Eyepiece Magnification Description
Availability Pricing
Meiji Techno Microscope Camera Adaptor, C-Mount, 0.3X, for 1/3" Chip Camera WW-48400-63 Mfr # MA151/35/03
COMPARE
0.3 Microscope Camera Adaptor, C-Mount, 0.3X, for 1/3\" Chip Camera
₹57,900.00INR / Each
Meiji Techno Microscope Camera Adaptor, C-Mount, No Lens WW-48402-72 Mfr # MA151/5N
COMPARE
None Microscope Camera Adaptor, C-Mount, No Lens
₹35,200.00INR / Each
Meiji Techno Microscope Camera Adaptor, C-Mount, 2.5X, for Canon 35 mm Camera WW-48402-81 Mfr # MA150/50+MA512+T2-1
COMPARE
2.5 Microscope Camera Adaptor, C-Mount, 2.5X, for Canon 35 mm Camera
₹73,400.00INR / Each
Meiji Techno Microscope Camera Adaptor, C-Mount, 2.5X, for Canon EOS 35 mm Camera WW-48402-82 Mfr # MA150/50+MA512+T2-9
COMPARE
2.5 Microscope Camera Adaptor, C-Mount, 2.5X, for Canon EOS 35 mm Camera
₹73,400.00INR / Each