McMillan Flow Flowmeter Accessories

Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories
  • Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
7,400.00 - ₹39,500.00INR / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
McMillan Flow 100-17T Ryton and Brass Sensors Cable assembly, 30" (0.9 m) L WW-32704-52 Mfr # 100-17T
COMPARE
Ryton and Brass Sensors Cable assembly, 30" (0.9 m) L
₹12,800.00INR / Each
McMillan Flow Power supply/output cable (0 to 5 VDC), 115 VAC. WW-32707-50 Mfr # A-115VAC
COMPARE
Power supply/output cable (0 to 5 VDC), 115 VAC.
₹32,900.00INR / Each
McMillan Flow Power supply/output cable, 0 to 5 VDC; 230 VAC WW-32707-55 Mfr # A-230VAC
COMPARE
Power supply/output cable (0 to 5 VDC), 230 VAC
₹31,900.00INR / Each
McMillan Flow Power supply/input/output cable (0 to 5 VDC), 115 VAC WW-32708-50 Mfr # C-115VAC
COMPARE
Power supply/input/output cable (0 to 5 VDC), 115 VAC
₹39,300.00INR / Each
McMillan Flow Power supply/output cable, 0 to 5 VDC, 230 VAC WW-32708-55 Mfr # C-230VAC
COMPARE
Power supply/output cable (0 to 5 VDC), 230 VAC
₹39,500.00INR / Each
McMillan Flow 110-00-17 Accessory Base plate for RYTON flowmeters WW-32709-90 Mfr # 110-00-17
COMPARE
Accessory Base plate for RYTON flowmeters
₹7,400.00INR / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat