Masterflex® Variable-Area Flowmeter Valve Cartridges
Control your flow with the addition of a valve
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than mL/min, convert them to mL/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.
8,200.00 - ₹26,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Material Housing Max Flow Rate (mL/min) Description
Availability Pricing
COMPARE
316 Stainless Steel 20000 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Low Flow
₹10,900.00INR / Each
COMPARE
316 Stainless Steel 60000 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Medium Flow
₹10,800.00INR / Each
COMPARE
316 Stainless Steel 60001 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, High Flow
₹10,800.00INR / Each
COMPARE
Aluminum 60000 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Medium Flow
₹8,200.00INR / Each
COMPARE
Aluminum 20000 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Low Flow
₹8,200.00INR / Each
COMPARE
Aluminum 600001 Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, High Flow
₹8,200.00INR / Each
COMPARE
Aluminum 1000 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 3
₹14,000.00INR / Each
COMPARE
Aluminum 2500 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 4
₹14,000.00INR / Each
COMPARE
Aluminum 6200 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 5
₹14,000.00INR / Each
COMPARE
Aluminum 6201 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 6
₹14,000.00INR / Each
COMPARE
316 Stainless Steel 1000 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 3
₹26,000.00INR / Each
COMPARE
316 Stainless Steel 2500 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 4
₹26,000.00INR / Each
COMPARE
316 Stainless Steel 6200 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 5
₹26,000.00INR / Each
COMPARE
316 Stainless Steel 6201 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 6
₹26,000.00INR / Each
COMPARE
Aluminum 200 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 1
₹14,000.00INR / Each
COMPARE
316 Stainless Steel 200 Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 1
₹26,000.00INR / Each
Showing 1- 16 of 16