Masterflex® Transfer Tubing, Tygon® 2375
Won't allow particulate build-up
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
These transparent tubes are ideal for use with highly aggressive chemicals – acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. NSF 51 Standard compliant and manufactured according to GMP. Temperature range is -108 to 130°F (-78 to 54°C). Can be sterilized with ethylene oxide or gamma irradiation.
17,400.00 - ₹108,000.00INR / Pkg of 1
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
33 1/16 1/8
₹17,400.00INR / Pkg of 1
COMPARE
55 1/16 3/16
₹17,500.00INR / Pkg of 1
COMPARE
22 3/32 5/32
₹18,100.00INR / Pkg of 1
COMPARE
33 1/8 1/4
₹26,500.00INR / Pkg of 1
COMPARE
16 5/32 7/32
₹22,600.00INR / Pkg of 1
COMPARE
22 3/16 5/16
₹28,300.00INR / Pkg of 1
COMPARE
19 1/4 3/8
₹31,200.00INR / Pkg of 1
COMPARE
16 5/16 7/16
₹36,900.00INR / Pkg of 1
COMPARE
14 3/8 1/2
₹39,500.00INR / Pkg of 1
COMPARE
24 3/8 5/8
₹72,000.00INR / Pkg of 1
COMPARE
19 1/2 3/4
₹83,800.00INR / Pkg of 1
COMPARE
16 5/8 7/8
₹104,000.00INR / Pkg of 1
COMPARE
14 3/4 1
₹108,000.00INR / Pkg of 1
COMPARE
15 1 1
₹98,100.00INR / Pkg of 1
Showing 1- 14 of 14