Masterflex® Transfer Tubing, Polytetrafluoroethylene (PTFE)
Won't crack under heat or pressure
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Certification: Resin is FDA-compliant (21 CFR 177.1550)
Temperature range: -400 to 500°F (-240 to 260°C)
Sterilize: Autoclave, ethylene oxide, or dry heat.
11,500.00 - ₹142,000.00INR / Pkg of 1
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
370 1/32 1/16
₹13,300.00INR / Pkg of 1
COMPARE
370 1/16 1/8
₹11,500.00INR / Pkg of 1
COMPARE
294 3/32 5/32
₹21,800.00INR / Pkg of 1
COMPARE
240 1/8 3/16
₹19,800.00INR / Pkg of 1
COMPARE
375 1/8 1/4
₹35,000.00INR / Pkg of 1
COMPARE
294 3/16 5/16
₹50,600.00INR / Pkg of 1
COMPARE
137 1/4 5/16
₹29,300.00INR / Pkg of 1
COMPARE
240 1/4 3/8
₹61,500.00INR / Pkg of 1
COMPARE
143 19/64 3/8
₹49,900.00INR / Pkg of 1
COMPARE
203 5/16 7/16
₹60,800.00INR / Pkg of 1
COMPARE
175 3/8 1/2
₹64,900.00INR / Pkg of 1
COMPARE
137 2/5 1/2
₹56,900.00INR / Pkg of 1
COMPARE
83 7/16 1/2
₹39,300.00INR / Pkg of 1
COMPARE
73 1/2 9/16
₹51,200.00INR / Pkg of 1
COMPARE
137 1/2 5/8
₹84,100.00INR / Pkg of 1
COMPARE
66 9/16 5/8
₹56,100.00INR / Pkg of 1
COMPARE
59 5/8 11/16
₹60,000.00INR / Pkg of 1
COMPARE
113 5/8 3/4
₹142,000.00INR / Pkg of 1
COMPARE
54 11/16 3/4
₹66,300.00INR / Pkg of 1
COMPARE
96 3/4 7/8
₹138,000.00INR / Pkg of 1
Showing 1 - 20 of 24