Masterflex® Transfer Tubing, Microbore
Ideal of extreme temperatures and harsh chemical environments
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. Temperature range is –400 to 500°F (–240 to 260°C); melts at 620°F (327°C). Tensile strength is 2500 to 4000 psi. PTFE tubing resin is UL-recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.
4,500.00 - ₹22,800.00INR / Roll of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Description
Availability Pricing
COMPARE
1/75 1/32 Microbore PTFE Tubing, 0.012"ID x 0.030"OD, 100 ft/roll
₹4,500.00INR / Roll of 1
COMPARE
1/50 3/64 Microbore PTFE Tubing, 0.022"ID x 0.042"OD, 100 ft/roll
₹5,100.00INR / Roll of 1
COMPARE
1/32 1/18 Microbore PTFE Tubing, 0.032"ID x 0.056"OD, 100 ft/roll
₹7,500.00INR / Roll of 1
COMPARE
3/64 1/15 Microbore PTFE Tubing, 0.042"ID x 0.066"OD, 100 ft/roll
₹6,600.00INR / Roll of 1
COMPARE
1/20 5/64 Microbore PTFE Tubing, 0.053"ID x 0.076"OD, 100 ft/roll
₹9,100.00INR / Roll of 1
COMPARE
Microbore PTFE Tubing Assortment Pack, 5 x 100 Ft (500 Ft total)
₹22,800.00INR / Pkg of 1