Masterflex® Transfer Tubing, Adapta Flex™
Maintain flexibility at high pressure
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Outstanding resistance to heat and ozone
Masterflex® Adapta Flex™ transfer tubing is ideal for general purpose applications requiring maximum flexibility. The EPDM tube and tube cover along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class C limited oil resistance certifications.
17,900.00 - ₹38,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 1/4 15/32
₹18,500.00INR / Each
COMPARE
100 3/8 21/32
₹29,900.00INR / Each
COMPARE
100 1/2 13/16
₹21,900.00INR / Each
COMPARE
50 5/8 59/64
₹17,900.00INR / Each
COMPARE
50 3/4 1 5/64
₹21,000.00INR / Each
COMPARE
50 1 1 25/64
₹38,400.00INR / Each