Masterflex® Spring-Loaded Direct-Reading, Variable-Area Flowmeters for Air/Water/N2, Polysulfone Housing, Dual Scales
Perfect low-cost durable process meter
  • Spring-loaded indicator allows inline mounting in any position
  • Nonreactive wetted parts are ideal for applications requiring a high level of purity
  • Direct reading—has English and metric scales
Spring-loaded, no-float design means these versatile, economical meters do not depend on gravity for accurate readings. Mount in any position in your flow line. Straight pipe runs are not required before or after the flowmeter. Pressure drop is only 4 psi full scale.
Flowmeters feature a knife-edged piston for easy reading which remains visible even through cloudy liquids. Working parts are easily removed for maintenance. Ideal for general water and air applications, including process water and cooling water lines. Flowmeters feature direct reading scales for water in GPM and LPM, as well as for air (at 90 psi) in scfmand SLPM.
46,700.00 - ₹123,000.00INR / Each
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (GPM) Process Connection (NPT(F))
Availability Pricing
COMPARE
Brass 5
₹48,300.00INR / Each
COMPARE
Brass 10
₹46,900.00INR / Each
COMPARE
Brass 10
₹47,400.00INR / Each
COMPARE
Brass 15
₹48,700.00INR / Each
COMPARE
Brass 15
₹47,600.00INR / Each
COMPARE
Brass 20
₹70,900.00INR / Each
COMPARE
Brass 30
₹72,600.00INR / Each
COMPARE
Brass 40
₹69,700.00INR / Each
COMPARE
Brass 50
₹71,600.00INR / Each
COMPARE
Brass 5
₹69,900.00INR / Each
COMPARE
Brass 15
₹70,500.00INR / Each
COMPARE
Brass 30
₹102,000.00INR / Each
COMPARE
Brass 40
₹102,000.00INR / Each
COMPARE
Brass 50
₹103,000.00INR / Each
COMPARE
316 SS 5
₹88,900.00INR / Each
COMPARE
316 SS 5
₹90,200.00INR / Each
COMPARE
316 SS 15
₹90,200.00INR / Each
COMPARE
316 SS 30
₹119,000.00INR / Each
COMPARE
316 SS 40
₹120,000.00INR / Each
COMPARE
316 SS 50
₹120,000.00INR / Each
Showing 1 - 20 of 32