Masterflex® Sanitary Gaskets, Polymers
Ensure a tight seal
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Always in stock—most materials and configurations available to ship same day
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match your specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class IV and cytotoxicity criteria. Contact Cole-Parmer for other sizes or materials.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from -58 to 446°F (-50 to 230°C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
560.00 - ₹9,000.00INR / Pkg of 10
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Gasket Size (in) Description
Availability Pricing
Garlock 42MPE-050 EPDM Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk WW-16011-62 Mfr # 42MPE-050
COMPARE
EPDM 1/2 EPDM Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
₹1,100.00INR / Pkg of 10
Garlock 42MPU-050 Buna-N Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk WW-16011-63 Mfr # 42MPU-050
COMPARE
Buna-N 1/2 Buna-N Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
₹1,100.00INR / Pkg of 10
Garlock 42MPSFY-050 FKM Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk WW-16011-64 Mfr # 42MPSF-050
COMPARE
FKM 1/2 FKM Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
₹2,600.00INR / Pkg of 10
Garlock 42RXPX-050 Silicone Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk WW-16011-66 Mfr # 42RXPX-050
COMPARE
Silicon 1/2 Silicone Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
₹1,800.00INR / Pkg of 10
COMPARE
Buna-N 4 Buna-N Sanitary Gasket, Black, 4" Tri-Clamp; 1/Pk
₹560.00INR / Each
COMPARE
EPDM 4 EPDM Sanitary Gasket, 4" Tri-Clamp; 1/Pk
₹920.00INR / Each
COMPARE
Platinum-cured silicone 1 1/2 Sanitary Gasket, Silicone, 1 1/2", 10/pk
₹3,900.00INR / Pkg of 10
COMPARE
Platinum-cured silicone 2 Silicone Sanitary Gasket, 2" Tri-Clamp; 10/Pack
₹5,200.00INR / Pkg of 10
COMPARE
Platinum-cured silicone 2 1/2 Sanitary Gasket, Silicone, 2 1/2", 10/pk
₹5,700.00INR / Pkg of 10
COMPARE
Viton 3/4 Viton Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
₹7,700.00INR / Pkg of 10
COMPARE
Viton 1 Viton Sanitary Gasket, 1" Tri-Clamp; 10/Pack
₹8,400.00INR / Pkg of 10
COMPARE
Viton 2 1/2 Viton Sanitary Gasket, 2-1/2" Tri-Clamp; 10/Pack
₹9,000.00INR / Pkg of 10
COMPARE
Buna N 3/4 Buna-N Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
₹3,800.00INR / Pkg of 10
COMPARE
EPDM 1 1/2 EPDM Sanitary Gasket, 1-1/2" Tri-Clamp; 10/Pack
₹3,900.00INR / Pkg of 10
COMPARE
EPDM 2 EPDM Sanitary Gasket, 2" Tri-Clamp; 10/Pack
₹3,700.00INR / Pkg of 10
COMPARE
PTFE 3/4 PTFE Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
₹4,700.00INR / Pkg of 10
COMPARE
PTFE 1 PTFE Sanitary Gasket, 1" Tri-Clamp; 10/Pack
₹6,100.00INR / Pkg of 10
COMPARE
PTFE 1 1/2 PTFE Sanitary Gasket, 1-1/2" Tri-Clamp; 10/Pack
₹5,800.00INR / Pkg of 10
COMPARE
PTFE 2 PTFE Sanitary Gasket, 2" Tri-Clamp; 10/Pack
₹8,000.00INR / Pkg of 10
COMPARE
PTFE 2 1/2 PTFE Sanitary Gasket, 2-1/2" Tri-Clamp; 10/Pack
₹6,400.00INR / Pkg of 10
Showing 1 - 20 of 34