Masterflex® Quick-Disconnect Coupler Fittings, Hose Barbed, Straight
Provides easy single-handed disconnection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Construction from low-density polyethylene (LDPE).
6,600.00 - ₹11,400.00INR / Pkg of 20
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Scienceware 19731-0552 Barbed Reducing Quick-Connect Couplers, 3/16" x 1/4" ID, LDPE, 12/Pk WW-06289-10 Mfr # 19731-0552
COMPARE
3/16 1/4 LDPE
₹11,400.00INR / Pkg of 12
COMPARE
1/8 7/32 LDPE
₹9,500.00INR / Pkg of 20
COMPARE
3/16 5/16 LDPE
₹6,600.00INR / Pkg of 20
COMPARE
3/8 1/4 LDPE
₹6,800.00INR / Pkg of 20
COMPARE
3/8 7/16 LDPE
₹7,800.00INR / Pkg of 20