Masterflex® NSF-Approved Turbine Flowmeters
Designed for low-viscosity flow measurement in water and beverage applications
  • NSF Standard 61 listed—ideal for use with water sampling and processing applications
  • Designed with 3/8” NPT(M) process connections
  • Suitable for use with 1.8 to 16 cSt liquids
  • Features an accuracy of ±2% of reading
Designed for low-flow OEM and pilot applications, these turbine flowmeters are highly accurate and repeatable and feature a Hall effect sensor for superior accuracy. The sensor’s standard power and output specifications make it easy to retrofit existing controllers and output to a PLC, recorder, or panel meter. The 316 stainless steel shaft coupled with Delrin® acetal bearings allows for accurate measurements during quick dispensing cycles. These low-cost units meet the requirements of NSF Standard 61, making them ideal for water or beverage dispensing applications or any application with water-based liquids.
33,700.00 - ₹42,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Description
Availability Pricing
COMPARE
2 0.2 NSF-Approved Turbine Flowmeter, 0.2 to 2 GPM, 3/8" NPT(M)
₹42,700.00INR / Each
COMPARE
4 0.4 NSF-Approved Turbine Flowmeter, 0.4 to 4 GPM, 3/8" NPT(M)
₹33,700.00INR / Each