Masterflex L/S Variable-Speed Console Drives
Compact and cost effective for general fluid transfer applications
  • Flow range: 0.42 to 2900 mL/min
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Drives accept nine different pump head types: Standard, Easy-Load®, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE tubing, and PTFE-diaphragm. Drives 07555-00 and -05 are not recommended for use with the PTFE-tubing and PTFE-diaphragm pump heads.
122,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max RPM (rpm) Power (VAC)
Availability Pricing
COMPARE
2900 600 90 to 130
₹122,000.00INR / Each
COMPARE
2900 600 180 to 260
₹122,000.00INR / Each
COMPARE
1130 200 90 to 130
₹122,000.00INR / Each
COMPARE
1130 200 180 to 260
₹122,000.00INR / Each