Masterflex L/S Pump Systems with MasterflexLive®
Cloud-enabled pump systems for secure, real-time monitoring and control of pump status and performance
  • MasterflexLive® is a secure, cloud-enabled platform for control and monitoring of Masterflex pumps
  • Get real-time control of all pump parameters – including speed, flow rate, dispense volume, and more
  • Push notifications alert you to operating conditions and error messages
  • Monitor and adjust critical processes running 24/7 without the need for onsite personnel
  • Internet connection via standard Ethernet (RJ45) jack on rear of drive
  • Flow rate 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually
  • Open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
559,000.00 - ₹793,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Remote I/O Housing Material Max Flow Rate Per Channel (mL/min)
Availability Pricing
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure ABS Plastic and Aluminum 480
₹559,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure ABS Plastic and Aluminum 1700
₹559,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure ABS Plastic and Aluminum 80
₹559,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure ABS Plastic and Aluminum 280
₹559,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure Stainless Steel 480
₹792,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure Stainless Steel 1700
₹793,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure Powder-Coat Steel 480
₹699,000.00INR / Each
COMPARE
Ethernet, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure Powder-Coat Steel 1700
₹700,000.00INR / Each
Showing 1- 8 of 8