Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone
Excellent biocompatibility that meets multiple classifications
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
13,100.00 - ₹24,600.00INR / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
₹13,100.00INR / Pkg of 1
COMPARE
1/16 14 130
₹17,200.00INR / Pkg of 1
COMPARE
1/8 16 480
₹15,900.00INR / Pkg of 1
COMPARE
1/4 17 1700
₹22,400.00INR / Pkg of 1
COMPARE
3/8 18 2300
₹24,600.00INR / Pkg of 1
COMPARE
3/16 25 1000
₹21,300.00INR / Pkg of 1