Masterflex® L/S® Precision Pump Tubing, PharMed® BPT
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
22,800.00 - ₹37,700.00INR / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
₹22,800.00INR / Pkg of 1
COMPARE
1/16 14 130
₹29,200.00INR / Pkg of 1
COMPARE
1/8 16 480
₹29,200.00INR / Pkg of 1
COMPARE
1/4 17 1700
₹36,700.00INR / Pkg of 1
COMPARE
3/8 18 2300
₹37,700.00INR / Pkg of 1
COMPARE
3/16 25 1000
₹30,700.00INR / Pkg of 1