Masterflex® Inert In-Line Check Valves
No maintenance required!
  • Ideal for high-purity, low pressure
  • No metal components — inert flow path
  • No back flow leakage with back pressure of 0 to 100 psi
  • 1/4-28 UNF threaded connections
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.
10,200.00 - ₹29,100.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Breaking Pressure (PSI) Cv Wetted Materials
Availability Pricing
COMPARE
0.99 0.014 PEEK/EPDM
₹10,500.00INR / Each
COMPARE
0.23 0.014 PEEK
₹28,100.00INR / Each
COMPARE
0.99 0.014 PEEK/EPDM
₹10,700.00INR / Each
COMPARE
0.23 0.014 PEEK
₹29,100.00INR / Each
COMPARE
0.99 0.014 PEEK/EPDM
₹10,200.00INR / Each
COMPARE
0.23 0.014 PEEK
₹25,800.00INR / Each