Masterflex® I/P® High-Performance Precision Pump Tubing, Platinum-Cured Silicone
Reduce the risk of particulate entrapment with smooth inner surface
  • Free Certificate of Compliance is available - request in cart
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • No leachable additives protects sensitive and costly fluids
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
44,400.00 - ₹225,000.00INR / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) I/P Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
3/8 70 8000
₹44,400.00INR / Pkg of 1
COMPARE
1/2 88 17000
₹56,700.00INR / Pkg of 1
COMPARE
5/8 89 19000
₹68,800.00INR / Pkg of 1
COMPARE
1/2 88 17000
₹225,000.00INR / Box