Masterflex® Entry-Level Touch-Screen Syringe Pumps
Accommodate up to two syringes in a cost-effective touch-screen syringe pump
  • Flow ranges from 0.54 pL up to 88.8 mL/min
  • Full-color touch screen
  • Daisy chain multiple pumps using the integrated RS-485
  • Use with any manufacturer syringe—glass, stainless steel, or plastic—from 0.5 ìL to 60 mL
These entry-level pumps offer features normally found in more expensive models. An intuitive touch-screen display lets you see all operating parameters and easily create and recall configurations. Front-panel LED light indicates when pump is operating—vie
387,000.00 - ₹573,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (picoL/min) Number of Syringes
Availability Pricing
COMPARE
88.28 1.28 1
₹387,000.00INR / Each
COMPARE
88.28 1.28 1
₹496,000.00INR / Each
COMPARE
88.28 1.28 2
₹471,000.00INR / Each
COMPARE
25.99 1.28 2
₹500,000.00INR / Each
COMPARE
11.7 0.54 2
₹499,000.00INR / Each
COMPARE
3.818 3.66 1
₹573,000.00INR / Each