Masterflex® Ball Valves, PTFE
PTFE wetted parts allow for maximum chemical resistance
  • Ideal for use in semiconductor or pharmaceutical manufacturing and chemical processing
  • Bidirectional 1/4-turn from OFF to ON
  • PTFE and polypropylene (PP) body construction
  • Perfect for use in harsh environments
176,000.00 - ₹274,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Max Pressure (PSI) Port Size (NPT(F))
Availability Pricing
COMPARE
6.0 80 0.5
₹176,000.00INR / Each
COMPARE
15.1 60 0.75
₹264,000.00INR / Each
COMPARE
29.5 60 1
₹274,000.00INR / Each