Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Plus Platinum-Cured Silicone
Ensure optimum performance from your Masterflex® pump
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Optically inspected, factory tested
  • Ensures flow rates are accurate and promotes longer service life
  • Seven different tubing formulations—choose tubing based on chemical compatibility, life, and pressure or vacuum needs
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
65,900.00 - ₹94,100.00INR / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
B/T Masterflex tubing size Length Per Pack (ft) Tubing ID (mm)
Availability Pricing
COMPARE
92 10 25.4
₹74,500.00INR / Pkg of 1
COMPARE
87 10 12.7
₹65,900.00INR / Pkg of 1
COMPARE
91 10 19
₹94,100.00INR / Pkg of 1