Marsh Bellofram Type 78 High-Flow Regulators
Designed for applications that require high flow and accurate pressure control
  • Rolling diaphragm provides low-friction, long life service
With a maximum supply pressure of up to 400 psi (27.6 bar) and output ranges up to 125 psi (8.6 bar), these pressure regulators can achieve flow rates of well over 200 SCFM. Once set to a desired output pressure, these direct-acting diaphragm-operated regulators maintain the setting permanently. The rugged die-cast zinc and aluminum housings are pressure tested to assure safe operation, and are designed to withstand harsh and abusive environments. Can be panel or pipe mounted.
55,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Exhaust Flow Rate (SCFM) Max Supply Pressure (PSI) Supply Pressure Effect
Availability Pricing
Marsh Bellofram 960-337-000 High-Flow Regulator, 0 to 125 psi, 200 scfm WW-68825-98 Mfr # 960-337-000
COMPARE
400 < 0.35 psi per 100 psi
₹55,100.00INR / Each