Magnetic tubing closures for Specra/Por RC Membranes
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
Magnetic closures contain a fluorocarbon-coated magnetic bar, eliminating the need for conventional stir bars. This closure provides complete submersion and gentle rotation of the sample on a magnetic stirrer.
21,400.00INR / Pkg of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Color Tubing Sealing Width (mm)
Availability Pricing
Spectra Por 132766 RC Dialysis Membrane Tubing Closure-Magnetic, 75MM Red 2/pk WW-02899-29 Mfr # 132766
COMPARE
Red 75
₹21,400.00INR / Pkg of 2