MadgeTech 4 or 8 Channel Thermocouple or RTD Data Loggers
This is an all-in-one compact, portable, easy to use device that will measure and record up to thousands of measurements per channel. The storage medium is non-volatile, solid-state memory - providing maximum data security even if the battery becomes discharged. The device can be started and stopped directly from your computer and its small size allows it to fit almost anywhere. Simply plug it into an empty COM port and our user-friendly software does the rest.
FEATURES
  • Type J, K, T, E, R, S, B, N Thermocouple or 100 Ohm RTD models
  • Real-time operation
  • Low cost
  • User-friendly
  • Battery powered - user replaceable
  • Programmable start time
  • Small size
  • Non-volatile solid state memory
  • Readings selectable 1 every 5 sec to 1 every 12 hours
26,300.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech TC-K-TT-24-36SMP T/C-K Probe, 36in. With Mini-Plug PTFE Insulated -270/1370C WW-18002-89 Mfr # TC-K-TT-24-36SMP
COMPARE
₹26,300.00INR / Each