Low-Pressure Full Faceshield Supplied-Air Respirator Kits
Complete and convenient NIOSH-approved systems you can take any where
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient systems come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Full-Faceshield Systems give you the convenience of a supplied air system with the protection of an impact-resistant faceshield. High impact polycarbonate lens also features anti-fog and anti-scratch coating. Faceshield features a flip-up nylon visor with a polypropylene chin guard for added strength and durability. Cotton shroud keeps debris from falling down the back of the neck. Faceshield assembly meets ANSI Z87.1-2003 for impact resistance.
Approval: NIOSH (as a system)
358,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Allegro 9245-01 One Man Full Faceshield Supplied Air System with 50 foot Hose WW-81520-30 Mfr # 9245-01
COMPARE
One Man Full Faceshield Supplied Air System with 50 foot Hose
₹358,000.00INR / Each