This product is now discontinued, View Related Products
Lovibond TB250 Handheld Turbidity Meters
Allow easy turbidity measurement in both the field and the lab
  • Available in both EPA- and ISO-compliant models for regulatory monitoring
  • Simple-to-use four-button interface
  • Durable housing stands up to harsh field conditions
The TB250 portable turbidity meters are available in both US EPA (white light) or ISO (IR light) models. Power-efficient microcircuit design makes the meter ideal for field applications—yielding up to 5000 tests on four AA batteries! Durable lamps provide an estimated 7- to 10-year bulb life.
Auto-ranging function automatically selects the correct turbidity range for your sample and provides the best resolution possible. Integrated diagnostics ensure proper operation and reading accuracy. Calibration is simple with included sample kit with silicone oil for better reproducibility.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Description
Availability Pricing
COMPARE
1100 NTU 0.01 NTU Handheld Turbidity Meter, EPA compliant
Call For Price
Lovibond TB250-IR Handheld Turbidity Meter, ISO compliant WW-99599-92 Mfr # 194210
COMPARE
1100 NTU 0.01 NTU Handheld Turbidity Meter, ISO compliant