This product is now discontinued, View Related Products
Love Economical On/Off Temperature Controllers
Digital display and adjustable deadband for a very economical price
  • Powerful 16 A, SPST relay can run up to a ¾ hp compressor
  • Feature password protection and error messages
  • Include thermistor probe with 5-ft cable
These controllers are ideal for on/off control applications where a digital display and an adjustable deadband are needed over a limited temperature range. Use with packaging equipment, food cooling and chilling equipment, wine cellars, and more!
Each controller has a tactile front keypad that lets you program 12 different parameters, including set point, deadband, cycle time and ambient probe adjustment. Programmable password protection prevents accidental changes to settings. The bright red, 3-digit LED display is easy to read even in dim areas.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
Dwyer TS-13020 Economical Temperature Controller, 220 V, °F WW-93520-05 Mfr # TS-13020
COMPARE
302 -58 Economical Temperature Controller, 220 V, °F
Call For Price
Dwyer TS-13030 Economical Temperature Controller, 12 VDC, °F WW-93520-08 Mfr # TS-13030
COMPARE
302 -58 Economical Temperature Controller, 12 VDC, °F
Call For Price
Dwyer TS-13011 Economical Temperature Controller, 110 V, °C WW-93520-10 Mfr # TS-13011
COMPARE
Economical Temperature Controller, 110 V, °C
Call For Price
Dwyer TS-13031 Economical Temperature Controller, 12 VDC, °C WW-93520-18 Mfr # TS-13031
COMPARE
Economical Temperature Controller, 12 VDC, °C