This product is now discontinued, View Related Products
Love A-Series Temperature/Process Controllers
Fuzzy PID logic eliminates temperature overshoot
  • Universal controller accepts thermocouple, RTD, voltage and current input
  • Weatherproof NEMA 4X (IP66) housing
  • Models available with 16-segment ramp and soak
These self-tuning controllers offer many options to fit your application. Choose from 1/32-, 1/16-, 1/8- (vertical), and 1/4-DIN sizes. Models are available with SPST alarm relay. A set point modifier creates a temporary set point that allows the controller to recover quickly. Models with fuzzy logic are available in 1/32-, 1/16-, and 1/4-DIN sizes. Controllers feature a variety of output configurations: SPST relay, SPDT relay, SSR, pulse output, or 4 to 20 mA output. Quiet and reliable 4 to 20 mA output includes a regulated, isolated 24 V loop power supply. Models with output relays require snubber 89800-99 when connecting to an inductive load; order snubber below. Order probes separately. Call our Application Specialists for more information on these products or to special order a controller with other options.
Call For Price
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Dwyer 16A2030 1/16 DIN controllers with universal input and fuzzy logic, SPST relay WW-89802-00 Mfr # 16A2030
COMPARE
1/16
Call For Price
Dwyer 16A2020 1/16 DIN controllers with universal input and fuzzy logic, pulse WW-89802-02 Mfr # 16A2020
COMPARE
1/16
Call For Price
Dwyer 16A2120 1/16 DIN controllers with universal input and fuzzy logic, pulse and alarm WW-89802-12 Mfr # 16A2120
COMPARE
1/16
Call For Price
Dwyer 16A2111 1/16 DIN controllers with universal input and fuzzy logic, two SSR and alarm WW-89802-21 Mfr # 16A2111
COMPARE
1/16
Call For Price
Dwyer 16A2122 1/16 DIN controllers with universal input and fuzzy logic, two pulse outputs and alarm WW-89802-22 Mfr # 16A2122
COMPARE
1/16
Call For Price
Dwyer 16A2153 1/16 DIN controllers with universal input and fuzzy logic; 4-20 mA, SPST relay, and alarm WW-89802-34 Mfr # 16A2153
COMPARE
1/16
Call For Price
Dwyer 32A030 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, SPST relay WW-89802-40 Mfr # 32A030
COMPARE
1/32
Call For Price
Dwyer 32A020 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, pulse WW-89802-42 Mfr # 32A020
COMPARE
1/32
Call For Price
Dwyer 32A010 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, SSR WW-89802-44 Mfr # 32A010
COMPARE
1/32
Call For Price
Dwyer 32A050 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, 4-20mA WW-89802-46 Mfr # 32A050
COMPARE
1/32
Call For Price
Dwyer 32A133 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, SPST relay WW-89802-48 Mfr # 32A133
COMPARE
1/32
Call For Price
Dwyer 32A113 1/32 DIN controllers with universal input, fuzzy logic, and ramp and soak, SSR WW-89802-50 Mfr # 32A113
COMPARE
1/32
Call For Price
Dwyer 16A3030 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; SPST relay WW-89803-00 Mfr # 16A3030
COMPARE
1/16
Call For Price
Dwyer 16A3020 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; pulse WW-89803-02 Mfr # 16A3020
COMPARE
1/16
Call For Price
Dwyer 16A3120 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; pulse and alarm WW-89803-12 Mfr # 16A3120
COMPARE
1/16
Call For Price
Dwyer 16A3133 1/16-DIN Universal Input Temp Controller; Fuzzy/2 SPST/Alarm/Ramp/Soak WW-89803-20 Mfr # 16A3133
COMPARE
1/16
Call For Price
Dwyer 16A3111 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; two SSR and alarm WW-89803-21 Mfr # 16A3111
COMPARE
1/16
Call For Price
Dwyer 16A3122 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; two pulse outputs and WW-89803-22 Mfr # 16A3122
COMPARE
1/16
Call For Price
Dwyer 16A3153 1/16 DIN controllers with universal input, fuzzy logic, and ramp and soak; 4-20 mA, SPST relay, WW-89803-34 Mfr # 16A3153
COMPARE
1/16
Call For Price
Dwyer 85113-0 1/8 DIN vertical style controllers with thermocouple input; SPDT relay and alarm WW-89804-10 Mfr # 85113-0
COMPARE
1/8
Call For Price
Showing 1 - 20 of 27