This product is now discontinued, View Related Products
Lock Out/Tag Out; Danger: Do Not Operate - My Life Is On The Line
These tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” hole. Choose from three materials based on application.
Cardstock (PF) is a wood pulp-free, tear-resistant, waterproof material, and is 10-mil poly-based.
Plastic (RV) is a 15-mil rigid vinyl plastic material that is durable and will withstand tears, weathering, and various chemicals.
Laminate (HS) is a tough durable poly-tag material for abusive, dirty, and greasy environments. The poly-tag is sealed on both sides by thick rigid vinyl. The plastic and laminate tags also have grommet re
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
Accuform MLT600LCP Tag, Danger Do Not Operate My Life Is On The Line, 5 7/8" X 3 1/8", PF-Cardstock WW-40503-33 Mfr # MLT600LCP
COMPARE
Cardstock, Laminate
Call For Price
Accuform MLT600LPP Tag, Danger Do Not Operate My Life Is On The Line, 5 7/8" X 3 3/8", RV-Plastic WW-40503-34 Mfr # MLT600LPP
COMPARE
Plastic