Lock Out/Tag Out; Danger - Do Not Operate - Group Lockout
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
3,700.00INR / Pkg of 25
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Accuform MDT169CTP Tag, Danger Do Not Energize, Job, 5 7/8" X 3 1/8", Pf-cardstock, 25/pk WW-40503-27 Mfr # MDT169CTP
COMPARE
Polyolefin
₹3,700.00INR / Pkg of 25