This product is now discontinued, View Related Products
ACTIONI/Q® IQRL Power Distribution Bus
Provides Power to Action I/Q Signal Conditioning Modules
  • Reduces Wiring Time
  • Improves Reliability with Fewer Wire to Terminal Connections
  • Attaches Directly to the ActionI/Q Module
  • SnapLoc Plug-In Terminals
These are designed to power 4-wire transmitters in the Action I/Q family, specifically models Q1x6, and Q4x6.
The Action I/Q modules are designed to hold the I/QRail firmly in place and parallel to the DIN mounting rail. No additional mounting holes or cutouts are required for I/QRail installation. Action I/Q modules mount on standard TS32 (32mm) or TS35 (35mm) DIN rails which then support the complete installation.
Model 68478-36, IQRL-2002, powers 2 AC modules, and model 68479-05, IQRL-2004, powers 4 AC modules.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Input type Max Input Temperature (° F) Power Supply (VAC)
Availability Pricing
Eurotherm I/q Rail DC Pwr Dist Asy 4mod WW-10508-14 Mfr # IQRL-D004
COMPARE
Call For Price
Eurotherm-Action Instruments IQRL-2002 Rail AC Power Distribution Bus; 2 Modules WW-68478-36 Mfr # IQRL-2002
COMPARE
Isolation: 1800V DC, between power conductors 265
Call For Price
Eurotherm IQRL-D004 I/q Rail DC Pwr Dist Asy 4mod WW-68478-58 Mfr # IQRL-D004
COMPARE
Call For Price
Eurotherm IQRL-D008 I/q Rail DC Pwr Dist Asy 8mod WW-68478-59 Mfr # IQRL-D008
COMPARE
265
Call For Price
Eurotherm IQRL-2004 Rail AC Power Distribution Bus; 4 Modules WW-68479-05 Mfr # IQRL-2004
COMPARE
Isolation: 1800V DC, between power conductors 120/240