Lascar Electronics Compact USB Data Loggers, Temperature - Internal Sensor
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds — perfect for compliance monitoring
  • All data loggers come with software
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Lascar Electronics EL-USB-1 USB Temperature Data Logger, 16,000+ readings, 1 year battery life WW-23039-50 Mfr # EL-USB-1
COMPARE
176 ±2 0.1
Call For Price
Lascar Electronics EL-USB-LITE USB Temperature Data Logger, 4,000+ readings, 1 month battery life WW-23039-51 Mfr # EL-USB-LITE
COMPARE
122 ±4.5
Call For Price
Lascar Electronics EL-USB-1-LCD USB Temperature Data Logger with Display WW-23039-53 Mfr # EL-USB-1-LCD
COMPARE
176 ±2 1
Call For Price
Lascar Electronics EL-USB-1-PRO High Temperature Data Logger, 32,000+ readings, 3 year battery life WW-23039-66 Mfr # EL-USB-1-PRO
COMPARE
257 ±0.4 0.2