This product is now discontinued, View Related Products

Large Capacity Stainless Steel Ultrasonic Cleaners

Capacities up to 22 gallons accommodate your large or bulky items
  • Microprocessor-based controls ensure precise process management
These stainless steel cleaners easily handle a variety of industrial cleaning applications—from densely packed smaller parts to heavily soiled larger components. Choose a 25 kHz unit for aggressive cleaning; choose a 40 kHz unit for more gentle cleaning. Use the digital controls to set cleaning time from 1 to 99 minutes and temperature from ambient to 176degF (80degC).
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Frequency Output (Hz) Power (VAC)
Availability Pricing
Branson CPN-908-011 Large-Capacity Stainless Steel Ultrasonic Cleaner, 25 Hz, 10 gal; 208VAC WW-08995-00 Mfr # CPN-908-011
COMPARE
10 208
Call For Price
Branson CPN-908-012 Large-Capacity Stainless Steel Ultrasonic Cleaner, 40 Hz, 10 gal; 208VAC WW-08995-02 Mfr # CPN-908-012
COMPARE
10 208
Call For Price
Branson CPN-908-013 Large-Capacity Stainless Steel Ultrasonic Cleaner, 25 Hz, 10 gal; 230VAC WW-08995-04 Mfr # CPN-908-013
COMPARE
10 230
Call For Price
Branson CPN-908-014 Large-Capacity Stainless Steel Ultrasonic Cleaner, 40 Hz, 10 gal; 230VAC WW-08995-06 Mfr # CPN-908-014
COMPARE
10 230
Call For Price
Branson CPN-908-021 Large-Capacity Stainless Steel Ultrasonic Cleaner, 25 Hz, 21 gal; 208VAC WW-08995-08 Mfr # CPN-908-021
COMPARE
21 208
Call For Price
Branson CPN-908-022 Large-Capacity Stainless Steel Ultrasonic Cleaner, 40 Hz, 21 gal; 208VAC WW-08995-09 Mfr # CPN-908-022
COMPARE
21 208
Call For Price
Branson CPN-908-023 Large-Capacity Stainless Steel Ultrasonic Cleaner, 25 Hz, 21 gal; 230VAC WW-08995-12 Mfr # CPN-908-023
COMPARE
21 230
Call For Price
Branson CPN-908-024 Large-Capacity Stainless Steel Ultrasonic Cleaner, 40 Hz, 21 gal; 230VAC WW-08995-14 Mfr # CPN-908-024
COMPARE
21 230
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat