LaMotte Economical Soil Sampler
Basic soil sampling for an economical price!
  • Cut-away side permits easy visual inspection of the soil sample
This new soil sampler gives you quick, no frills core samples at an economical price. Sampler is made of galvanized steel with a tapered saw-toothed cutting edge. Soil sampler cuts a 1" dia x 10"L core. Samplers include 20 LDPE sample storage bags. Shpg wt 1.5 lb (0.68 kg).
13,300.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Core Diameter (cm) Length (cm) Material
Availability Pricing
LaMotte 1055/EP Handheld Soil Sampler; 10" Length, 1" Core Diameter 9902700 Mfr # 1055/EP
COMPARE
2.54 25.4 Galvanized Steel
₹13,300.00INR / Each