This product is now discontinued, View Related Products

Labomed Phase Contrast Microscopes

View transparent specimens in wastewater treatment, dairy, and water purification applications
  • Built-in ergonomic hand rests for focusing controls increases comfort during use
View transparent specimens such as living cells, tissue slices, and microorganisms, without having to use stains. All models have an extended base for stability, ergonomic hand rests for the coarse/fine focusing controls, and an Abbe condenser with rack and pinion movements for illumination control. Viewing heads have interpupillary adjustment to ease eyestrain and diopter adjustment on both eyepieces.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Head Configuration Objectives Stage Dimensions
Availability Pricing
Labomed 9135014 Basic Phase Contrast Microscope, Binocular WW-49402-11 Mfr # 9135014
COMPARE
Binocular Semi-plan phase and semi-plan brightfield 124 x 135 mm
Call For Price
Labomed 9135015 Basic Phase Contrast Microscope, Trinocular WW-49402-12 Mfr # 9135015
COMPARE
Trinocular Semi-plan phase and semi-plan brightfield 124 x 135 mm
Call For Price
Labomed 9126005 Advanced Phase Contrast Microscope, Binocular WW-49402-13 Mfr # 9126005
COMPARE
Binocular Plan phase 160 x 200 mm
Call For Price
Labomed 9126006 Advanced Phase Contrast Microscope, Trinocular WW-49402-14 Mfr # 9126006
COMPARE
Trinocular Plan phase 160 x 200 mm
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat