This product is now discontinued, View Related Products
Labconco Purifier Horizontal Clean Benches
Remove particles from room air to supply Class 100 air to your bench working area
  • Provides ISO Class 5 conditions and product protection
  • Two interior depths offered to match your application needs
  • Easy to clean, static-dissipative stainless steel work surface
Sterile horizontal laminar flow benches filter out particles as small as 0.3µm with 99.99% efficiency, providing ISO Class 5 air (fewer than 3520 particles, 0.5 µm or larger per cubic meter of air). Prefilters trap large particles, extending HEPA filter life. Differential pressure gauge displays pressure across the HEPA filter.
Benches are constructed of powder-coated steel exterior, safety glass side panels with utility ports, and 304 stainless steel work surface. Models with built-in 254 nm UV light feature retractable UV screen. Light timer provides timed UV light exposure (5, 10, 15, 30, 60, 120, 240 minutes and continuous settings). Benches include a built-in motor blower with vibration isolation pad. Other 230-volt plug types are available; contact an Application Specialist for details.
Note: Benches provide sample protection only. No personnel protection is provided. Do not use with biohazards, toxins or radionuclides.
Compliance: ETL (115 volt models), CE (230 volt models)
Accessories
Labconco Telescoping Base Stands — support Purifier® Horizontal Clean Benches
Stands feature telescoping legs so you can select stand height during installation. Height is adjustable to eight positions in 1” increments from 27.5” to 34.5”. Frame is durable 1.75” tubular, powder-coated steel with rear crossbar supports. ADA compliant. Fixed leveling feet.
Labconco Replacement Filters for Purifier® Horizontal Clean Benches
*38-39” W and 50-51” W clean benches require two prefilters, 74-75” wide clean benches require three prefilters, and 99-100” wide clean benches require four prefilters.
** 99-100” wide clean benches require two HEPA filters.
Call For Price
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Exterior Depth (in) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco Purifier 3 ft Horizontal Clean Bench, 34” D, 115V WW-09116-00 Mfr # 3889700
COMPARE
34 38 115
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, 34” D, UK WW-09116-02 Mfr # 3889705
COMPARE
34 38 230
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, 39” D, 115V WW-09116-04 Mfr # 3889701
COMPARE
39 38 115
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, 39” D, UK WW-09116-06 Mfr # 3889706
COMPARE
39 38 230
Call For Price
Labconco Purifier 4 ft Horizontal Clean Bench, 34" D/115V WW-09116-08 Mfr # 3888400
COMPARE
34 50 115
Call For Price
Labconco Purifier 4 ft Horizontal Clean Bench, 34" D/UK WW-09116-10 Mfr # 3888405
COMPARE
34 50 230
Call For Price
Labconco Purifier 4 ft Horizontal Clean Bench, 39" D/115V WW-09116-12 Mfr # 3888401
COMPARE
39 50 115
Call For Price
Labconco Purifier 4 ft Horizontal Clean Bench, 39" D/UK WW-09116-14 Mfr # 3888406
COMPARE
39 50 230
Call For Price
Labconco Purifier 6 ft Horizontal Clean Bench, 34" D/115V WW-09116-16 Mfr # 3873000
COMPARE
34 74 115
Call For Price
Labconco Purifier 6 ft Horizontal Clean Bench, 34" D/UK WW-09116-18 Mfr # 3873005
COMPARE
34 74 230
Call For Price
Labconco Purifier 6 ft Horizontal Clean Bench, 39" D/115V WW-09116-20 Mfr # 3873001
COMPARE
39 74 115
Call For Price
Labconco Purifier 6 ft Horizontal Clean Bench, 39" D/UK WW-09116-22 Mfr # 3873006
COMPARE
39 74 230
Call For Price
Labconco Purifier 8 ft Horizontal Clean Bench, 34" D/115V WW-09116-24 Mfr # 3818500
COMPARE
34 99 115
Call For Price
Labconco Purifier 8 ft Horizontal Clean Bench, 34" D/UK WW-09116-26 Mfr # 3818505
COMPARE
34 99 230
Call For Price
Labconco Purifier 8 ft Horizontal Clean Bench, 39" D/115V WW-09116-28 Mfr # 3818501
COMPARE
39 99 115
Call For Price
Labconco Purifier 8 ft Horizontal Clean Bench, 39" D/UK WW-09116-30 Mfr # 3818506
COMPARE
39 99 230
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, UV, 34” D/115V WW-09116-32 Mfr # 3889720
COMPARE
34 39 115
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, UV, 34” D/UK WW-09116-34 Mfr # 3889725
COMPARE
34 39 230
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, UV, 39” D/115V WW-09116-36 Mfr # 3889721
COMPARE
39 39 115
Call For Price
Labconco Purifier 3 ft Horizontal Clean Bench, UV, 39” D/UK WW-09116-38 Mfr # 3889726
COMPARE
39 39 230
Call For Price
Showing 1 - 20 of 32