This product is now discontinued, View Related Products
Labconco Protector Airo™ Ductless Fume Hoods
No infrastructure changes required
  • Compact size fits in tight spaces, interior rooms, and basements with low ceilings
  • Uses the Erlab® GFH filtration technology and Neutrodine® filters
  • Ergonomic sash is engineered to reduce turbulence and safely stow away power cords
These compact fume hoods require as little as eight feet of height for installation, perfect for small spaces. The high-capacity Neutrodine® filters (order separately, required) scrub a wide variety of chemicals out of the air, and onboard sensors detect the saturation point of the primary filter and alert you with audible and visual alarms while the secondary filter is still functional, allowing filter exchanges at your convenience. HEPA filter is H14 type, 99.995% efficient (order separately, optional)
Air foil, sash handle, and sash tracks pull air toward the interior and reduce turbulence. Slots on either side of the air foil allow sash to be closed completely while electrical cords are plugged into the GFCI duplex outlet. Hoods with GFCI duplex are rated at 20 A.
Control access to hood operation with radio frequency identification (RFID) cards. This RFID technology also prevents re-installation of used filters by scanning filter casing serial numbers.
Note: Order fume hood, filters, work surfaces, and base cabinets or stands separately.
Call For Price
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 115 V SKU 2861457 WW-28614-57 Mfr # 184300000
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 115 V SKU 2861458 WW-28614-58 Mfr # 184300001
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 1 Receptacle, 115 V WW-28614-59 Mfr # 184300002
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 1 Receptacle, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3'W x 54" H, 1 Receptacle WW-28614-60 Mfr # 184300003
COMPARE
Ductless Fume Hood, 3'W x 54\" H, 1 Receptacle
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54 "H, 230 V WW-28614-61 Mfr # 184300020
COMPARE
Ductless Fume Hood, 3' W x 54 \"H, 230 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 230 V SKU 2861462 WW-28614-62 Mfr # 184300021
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 230 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 115 V SKU 2861463 WW-28614-63 Mfr # 184310000
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 115 V SKU 2861464 WW-28614-64 Mfr # 184310001
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 1 Receptacle, 115 V WW-28614-65 Mfr # 184310002
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 1 Receptacle, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 1 Receptacle, 115                                                                        V WW-28614-66 Mfr # 184310003
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 1 Receptacle, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 230 V WW-28614-67 Mfr # 184310020
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 230 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 3' W x 54" H, 2 Fixtures, 230 V SKU 2861468 WW-28614-68 Mfr # 184310021
COMPARE
Ductless Fume Hood, 3' W x 54\" H, 2 Fixtures, 230 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W x 54" H, 115 V WW-28614-69 Mfr # 184410000
COMPARE
Ductless Fume Hood, 4' W x 54\" H, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W x 54" H, 2 Fixtures, 115 V WW-28614-71 Mfr # 184410001
COMPARE
Ductless Fume Hood, 4' W x 54\" H, 2 Fixtures, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W x 54" H, 2 Fixtures, 1 Receptacle, 115 V WW-28614-72 Mfr # 184410002
COMPARE
Ductless Fume Hood, 4' W x 54\" H, 2 Fixtures, 1 Receptacle, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W X 54" H, 1 Receptacle, 115 V WW-28614-73 Mfr # 184410003
COMPARE
Ductless Fume Hood, 4' W X 54\" H, 1 Receptacle, 115 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W x 54" H, 230 V WW-28614-74 Mfr # 184410020
COMPARE
Ductless Fume Hood, 4' W x 54\" H, 230 V
Call For Price
Labconco Protector Airo Ductless Fume Hood, 4' W x 54 "H, 2 Fixtures, 230 V WW-28614-76 Mfr # 184410021
COMPARE
Ductless Fume Hood, 4' W x 54 \"H, 2 Fixtures, 230 V
Call For Price
Showing 1- 18 of 18