This product is now discontinued, View Related Products
Labconco FreeZone Legacy 2.5-Liter Benchtop Freeze Dryers
Perfect for processing light sample loads in a limited bench space.
  • LCD display system
  • Audible and visual alarm for abnormal system event
  • Moisture sensor prevents refrigeration or vacuum start-up when moisture in the drain line is detected
  • Vaccum break valve bleeds air into system when power is shut off
These FreeZone Legacy freeze dryers have an upright stainless steel collector coil (PTFE-coated collector on 03338-05 and -15) with clear acrylic lid that holds 2.5 liters of ice before defrosting. The -50ºC (-58ºF) dryers have a single 1/3 hp HCFC/CFC-free refrigeration system which are suitable for aqueous samples. The dryers are easy to operate with push button control and LCD display. User configured to display vacuum (mBar, Torr, or Pa), and temperature (ºC or ºF). Other displays include total hours of refrigeration system operation, time since refrigeration system was serviced, total hours of vacuum pump operation, and hours since vacuum pump was serviced. Unit also has LED of vacuum and temperature "waves" for display of relative system vacuum and collector temperature.
Alarm feature flashes and beeps if any abnormal system events have occured, including: power failure, improper line voltage supply, collector temperature above -40°C, vacuum pump service needed, and moiture in collector. RS-232 port transmits data at user supplied intervals of 10, 30, 60, 300, or 600 seconds. Other features include; 3" diameter port for connection of drying accessories (sold separately), moisture sensor, vacuum break valve, retractable 9" collector drain hose, and electrical receptacle for vacuum pump connection.
All models require (not included) vacuum pump with a displacement of at leat 98 liters per minute, 0.002 mBar ultimate pressure and fitting suitable for 3/4" ID vacuum hose, drying accessories, and freeze dry glassware.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Power (VAC) Description
Availability Pricing
Labconco FreeZone FreeZone Legacy 2.5 L Benchtop Freeze Dry System, -50° with SS interior, 115V WW-03338-00 Mfr # 7670520
COMPARE
2.5 115 Freeze Dryer with SS Collector Coil, 115V, North American Plug
Call For Price
Labconco FreeZone FreeZone Legacy 2.5 L Benchtop Freeze Dry System, -50° with PTFE interior, 115V WW-03338-05 Mfr # 7670521
COMPARE
2.5 115 Freeze Dryer with PTFE Collector Coil, 115V, North American Plug
Call For Price
Labconco FreeZone FreeZone Legacy 2.5 L Benchtop Freeze Dry System, -50° with SS interior, 230V WW-03338-10 Mfr # 7670530
COMPARE
2.5 220 Freeze Dryer with SS Collector Coil, 230V, EU Plug
Call For Price
Labconco FreeZone FreeZone Legacy 2.5 L Benchtop Freeze Dry System, -50° with PTFE interior, 230v WW-03338-15 Mfr # 7670531
COMPARE
2.5 220 Freeze Dryer with PTFE Collector Coil, 230V, EU Plug
Call For Price
Labconco FreeZone 7670540 2.5 L Benchtop Freeze Dry System, -54° with SS interior, 208V WW-03338-20 Mfr # 7670540
COMPARE
4.5 115 2.5 L Benchtop Freeze Dry System, -54° with SS interior, 208V
Call For Price
Labconco FreeZone 7670541 2.5 L Benchtop Freeze Dry System, -54° with PTFE interior, 115V WW-03338-25 Mfr # 7670541
COMPARE
12 220 2.5 L Benchtop Freeze Dry System, -54° with PTFE interior, 115V