Labconco® CentriVap® Mobile System
Space-saving console combines the concentrator and cold trap components into one to conveniently transport
  • Small footprint makes it easy to move from lab to lab
  • Protect your vacuum pump—built-in cold trap collects up to 4 L of evaporated liquid
  • Recover as small as a few microliters using centrifugal force, vacuum, and heat
  • Create up to nine stored programs that can be altered while in progress
The Labconco® CentriVap® mobile system includes a vacuum concentrator, -55°C cold trap, and 12-13 mm rotor. Casters make it easy to transport from lab to lab. A glass lid allows you to easily view samples while offering protection against etching caused by solvents. The cold trap holds up to four liters of liquid to protect the vacuum concentrator from corrosion. A cutout in the front panel provides easy trap monitoring while allowing space for a vacuum pump; purchase separately.
The included 12-13 mm rotor accommodates up to 108 tubes. The 12” diameter chamber allows you to easily accommodate other rotors to hold up to 148 samples for greater throughput; order separately.
Control time and temperature using the easy-to-use keyboard. Up to nine programs can be quickly set and recalled and altered while in progress using the LCD. Define the temperature from -55 to 100°C in one degree increments—reach -55°C in less than 15 minutes.
Optional heat boost features a secondary 300 W heater on the sidewall of the chamber near the top to increase the evaporation speed.
Purchase vacuum pump and glassware separately to meet your application needs.
3,854,000.00 - ₹3,983,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Temperature (° C) Power (VAC)
Availability Pricing
Labconco 7812010 CentriVap® Mobile System; 115 VAC, 60 Hz WW-13040-87 Mfr # 7812010
COMPARE
1725 100 115
₹3,854,000.00INR / Each
Labconco 7812012 CentriVap® Mobile System; 230 VAC, 60 Hz WW-13040-88 Mfr # 7812012
COMPARE
1725 100 230
₹3,854,000.00INR / Each
Labconco 7812013 CentriVap® Mobile System, Heat Boost; 115 VAC, 60 Hz WW-13040-89 Mfr # 7812013
COMPARE
1725 100 115
₹3,983,000.00INR / Each
Labconco 7812015 CentriVap® Mobile System, Heat Boost; 230 VAC, 60 Hz WW-13040-90 Mfr # 7812015
COMPARE
1725 100 230
₹3,983,000.00INR / Each