Labconco CApture Portable Fuming System
Quickly fixes latent fingerprints at the crime scene
  • Unique collapsible design is easy to transport and set up
  • Uses microburst or traditional CA fuming methods
  • Protects the user from hazardous fumes without ducting
Bring this portable fuming system directly to the crime scene, saving valuable laboratory time and space. System combines all the components needed for fingerprint fuming in one compact, easy-to-transport unit. A microprocessor-controlled heater heats the CA adhesive (Super Glue®). The heater may be set in one degree increments between 120 to 350°F (49 to 77°C). An LCD displays the current or set-point temperature. A shatter-resistant, clear polycarbonate protective chamber, 8" W x 16" H x 8" D (20 x 41 x 20 cm), contains the fume as they react with the latent prints. A built-in exhaust blower pulls air from the chamber, through the filter and exhausts it into the room. A particulate/carbon exhaust filter traps smoke and CA fumes exhausted from the chamber.
Constructed of durable epoxy-coated steel with a stainless steel manifold to support evidence inside the chamber. A stainless steel hook, shelf and perforated tray are included. The CApture system runs on 115 VAC, 60 Hz, , 230 VAC, 50 Hz, or can be run on car battery when connected to a user-supplied inverter.
716,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco CApture Portable Fuming Station, 115 V WW-28616-08 Mfr # 3420000
COMPARE
19 10.75 115
₹716,000.00INR / Each