This product is now discontinued, View Related Products

Lab Companion Gravity Convection Ovens

Natural convection system is ideal for sterilization and drying applications requiring a gentle airflow
  • Dual-wall door opens to 180° and features double rack door handle for greater access to oven chamber
  • Rounded inner chamber corners offer easy cleaning and optimum air flow
  • Two 1.5'' diameter adjustable top-mounted ventilation slides control inner air and vapor circulation
Oven controller offers safety and programmable convenience features such as user-specified start and stop allowing you to perform other laboratory tasks. CLS (Custom Logical Safe)-Control system allows safety system to run independently of CPU performance controller — in the event of CPU failure, safety features are not compromised.
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Jeio Tech Lab Companion Gravity Convection Oven 1.8 cu.ft; 115VAC WW-52402-01 Mfr # AAH11136U
COMPARE
1.8 115
Call For Price
Jeio Tech Lab Companion Gravity Convection Oven 3.2 cu.ft; 115VAC WW-52402-02 Mfr # AAH11146U
COMPARE
3.2 115
Call For Price
Jeio Tech Lab Companion Gravity Convection Oven 4.8 cu.ft; 115VAC WW-52402-03 Mfr # AAH11156U
COMPARE
4.8 115
Call For Price
Jeio Tech Lab Companion Gravity Convection Oven 1.8 cu.ft; 230VAC Sku5240206 WW-52402-06 Mfr # AAH11135K
COMPARE
1.8 230
Call For Price
Jeio Tech Lab Companion Gravity Convection Oven 3.2 cu.ft; 230VAC WW-52402-07 Mfr # AAH11145K
COMPARE
3.2 230
Call For Price
Jeio Tech Lab Companion Gravity Convection Oven 4.8 cu.ft; 230VAC WW-52402-08 Mfr # AAH11155K
COMPARE
4.8 230
Call For Price
Jeio Tech Lab Companion AAH11136U Gravity Convection Oven 1.8 cu.ft; 115VAC WW-52402-71 Mfr # AAH11136U
COMPARE
2 115
Call For Price
Jeio Tech Lab Companion AAH11146U Gravity Convection Oven 3.2 cu.ft; 115VAC WW-52402-72 Mfr # AAH11146U
COMPARE
3 115
Call For Price
Jeio Tech Lab Companion AAH11156U Gravity Convection Oven 4.8 cu.ft; 115VAC WW-52402-73 Mfr # AAH11156U
COMPARE
5 115
Call For Price
Jeio Tech Lab Companion AAH11135K Gravity Convection Oven 1.8 cu.ft; 230VAC WW-52402-74 Mfr # AAH11135K
COMPARE
2 230
Call For Price
Lab Companion AAH11145K Gravity Convection Oven 3.2 cu.ft; 230VAC WW-52402-75 Mfr # AAH11145K
COMPARE
3 230
Call For Price
Lab Companion AAH11155K Gravity Convection Oven 4.8 cu.ft; 230VAC WW-52402-76 Mfr # AAH11155K
COMPARE
5 230
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat