Lab Companion Dual-Action Shakers
New user-friendly control panel is easy to read and intuitive to use
  • Dual-action shaking motion––select either orbital or reciprocating
  • Smooth start/stop function helps prevent spills from flasks or tubes
  • Platform stops where it starts, making repetitive tasks easy
  • Automatic shaking speed adjustment if an unbalanced or excessive workload is detected
Dual-action shakers have microprocessor PID control enabling reproducible results every time. The user-friendly control panel is easily read with a bright clear screen and the durable membrane touch switch and knob make setting and adjusting the speed simple and easy. Speed can be set and displayed in increments of 1, 5, or 10 depending on user preference.
Maintenance-free motor generates smooth and powerful shaking motion over a wide speed range and under heavy workloads. Built-in protection system from over-current, stalled platform, and audible/visible alarms for shaking speed deviation keep the unit worry free. RS-232 and USB ports provide option for external control and data collection.
Platform and accessories must be purchased separately.
461,000.00 - ₹592,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Platform Length (in) Platform Width (in) Power (VAC)
Availability Pricing
Jeio Tech Lab Companion OS-2000 Dual-Action Shaker, 20 to 500 rpm, 13.9" W × 15.3" L; 120 V WW-86578-26 Mfr # AAH3D1165U
COMPARE
10.2 12.6 120
₹461,000.00INR / Each
Jeio Tech Lab Companion OS-2000 Dual-Action Shaker, 20 to 500 rpm, 13.9" W × 15.3" L; 230 V WW-86578-27 Mfr # AAH3D1115K
COMPARE
10.2 12.6 230
₹461,000.00INR / Each
Jeio Tech Lab Companion OS-3000 Dual-Action Shaker, 20 to 500 rpm, 16.1" W × 20.5" D Platform; 120 V WW-86578-28 Mfr # AAH3D1265U
COMPARE
13.8 13.8 120
₹592,000.00INR / Each
Jeio Tech Lab Companion OS-3000 Dual-Action Shaker, 20 to 500 rpm, 16.1" W × 20.5" D Platform; 230 V WW-86578-29 Mfr # AAH3D1215K
COMPARE
13.8 13.8 230
₹592,000.00INR / Each