This product is now discontinued, View Related Products
Lab Companion Chamber-Style Incubating Shaker Kit
Bundled together for convenience and to save you money!
  • Platform returns to center position every time, making repetitive tasks easy to perform
  • Automatic speed adjustment keeps unit stable in case of an unbalanced load, uneven surface, or external shock
  • Prevent spills with smooth start and stop function
  • Stack up to two units to save valuable lab space
  • RS-232 port and USB ports for external control and data collection
These chamber-style incubating shakers have an intuitive touch-screen LCD controller with easy-to-use icons and logical menus. Controls include automatic run with set point memory after a power interruption, auto-tuning, three-point temperature calibration, and a three-step adjustable fan for uniform temperature distribution and recovery after door opening. Safety features include audible/visible alarm activated by shaking speed deviation, protection system against overcurrent or stalled platform, and three temperature monitoring systems.
Maintenance-free motor generates quiet, uniform, and powerful shaking motion for heavy workloads over a 10 to 500 rpm speed range. Large stainless steel chamber is corrosion resistant and features a built-in electrical outlet with cover. Two, adjustable-height shelves are included and allow for increased storage and static incubating of samples. Large window with LED lighting makes it easy to view samples without disrupting program.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Jeio Tech Lab Companion AAH23415U Complete Incubating Shaker Kit, 80 L, Universal; 120 VAC WW-25000-36 Mfr # AAH23415U
COMPARE
Complete Incubating Shaker Kit, 80 L, Universal; 120 VAC
Call For Price
Jeio Tech Lab Companion AAH23615U Complete Incubating Shaker Kit, 150 L, Universal; 120 VAC WW-25000-40 Mfr # AAH23615U
COMPARE
Complete Incubating Shaker Kit, 150 L, Universal; 120 VAC