This product is now discontinued, View Related Products
Kuntze Free Chlorine Disinfectant Analyzer Systems
Low-maintenance measurement and control of free chlorine
  • User-friendly, plain text menu guidance
  • Security of password control functions
  • Display of relay status and error messages
  • Two P/PI-controller with two relays
  • Integrated flow monitoring with Pt 100 temperature reading
  • Low-maintenance, automatic sensor cleaning on Krypton systems
  • Scalable, galvanically isolated 4 to 20 mA output
  • Invertible alarm output; permanent or pulse contact
Kuntze Krypton free chlorine systems provide low maintenance solutions for on-line disinfectant. These plug-and-play analyzers are designed for easy and accurate readings. The analyzer employs a unique metallic tip electrode with built-in 100 Pt temperature sensor that is able to withstand aggressive water environments where membrane tip electrodes experience failure.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Control Type Output Current
Availability Pricing
Kuntze K100 CL2 M-117 Free Chlorine Disinfectant Analyzer System; 110 VAC WW-27400-01 Mfr # 765444-CL2 M
COMPARE
On/Off 0-20mA
Call For Price
Kuntze K CL2- 117 Free Chlorine Analyzer System with Self Cleaning, 117 VAC WW-27400-03 Mfr # 70005200K
COMPARE
On/Off 0-20mA
Call For Price
Kuntze K100 CL2 M-117 Free Chlorine Disinfectant Analyzer System; 220 VAC WW-27400-07 Mfr # 765444-CL2 M
COMPARE
On/Off 0-20mA
Call For Price
Kuntze K CL2- 117 Free Chlorine Analyzer System with Self Cleaning, 220 VAC WW-27400-09 Mfr # 70005200K
COMPARE
On/Off 0-20mA