This product is now discontinued, View Related Products
Koehler Mechanically Refrigerated Cold Filter Plugging Point Bath
Cascade hermetic cooling system achieves temperatures down to –90°F (–68°C)
  • Conforms to ASTM D6371, IP309, and DIN 51428 specifications
Determine the low-temperature flow characteristics of biofuels, diesel fuels, and gas oils. Mechanically refrigerated bath uses cascade hermetic cooling to reach temperatures as low as –90°F (–68°C).
Bath consists of an insulated stainless steel tank and polished stainless steel cabinet. Insert the cold filter plugging point apparatus (sold separately) into the top plate of the bath. The vacuum system (sold separately; pump not included) has everything needed to connect a vacuum pump to the cold filter plugging point apparatus.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K45950 Cold Filter Plugging Point Bath, 115 VAC, 60 Hz WW-59876-90 Mfr # K45950
COMPARE
115
Call For Price
Koehler K45995 Cold Filter Plugging Point Bath, 220 to 240 VAC, 50 Hz WW-59876-91 Mfr # K45995
COMPARE
220