This product is now discontinued, View Related Products
Koehler Copper Corrosion Test Bath System
Microprocessor temperature controller for stability
  • Complies with ASTM D130 specifications
Determine the corrosiveness to copper of petroleum-based fuels and biodiesel by immersing a copper strip in the test sample at an elevated temperature. Compare the tarnish color of the strip to a ASTM Copper Strip Corrosion Standard (sold separately) to assess the relative corrosiveness level.
The constant-temperature bath features a digital temperature controller and a 750-watt heater; maintains a temperature stability of ±2°F (1°C). Overtemperature control interrupts power to the heater if programmed temperature is reached. Built-in support rack holds up to 16 test tubes.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K25330 Copper Corrosion Test Bath, 115 VAC WW-59876-30 Mfr # K25330
COMPARE
115
Call For Price
Koehler K25339 Copper Corrosion Test Bath, 230 VAC WW-59876-31 Mfr # K25339
COMPARE
230